missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat CSP alpha (aa 180-198) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP287
This item is not returnable.
View return policy
Description
Rat CSP alpha (residues 180-198) Synthetic Peptide, PEP-287, from Invitrogen.
PEP-287 is a 19 amino acid synthetic peptide whose sequence is derived from the rat CSP protein. The sequence of this peptide is (amino to carboxy terminus): E (180)-T-T-Q-L-T-A-D-S-H-P-S-Y-H-T-D-G-F-N (198)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, Cysteine String Protein Alpha, catalog # PA1-776. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
79130 | |
purified | |
Rat CSP alpha (aa 180-198) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
P60905 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Dnajc5 | |
No preservative | |
50 μg | |
E(180) - T - T - Q - L - T - A - D - S - H - P - S - Y - H - T - D - G - F - N(198) |