Learn More
OHAUS™ Ranger™ Count 4000
Shop All Ohaus Products
Click to view available options
Capacity:
3.00 kg
6.00 kg
15.00 kg
30.00 kg
Model:
RC41M15
RC41M15-M
RC41M3
RC41M3-M
RC41M30-M
RC41M6-M
Unit Size:
Each
Description
- Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
- Accurate weighing and counting performance is assured with 1:1,500,000 internal counting resolution and auto-optimization software that reduces errors.
- Easy-to-operate with SmarText™ software, a centrally located count display and dedicated Sample and Tare keys.
Specifications
Specifications
| Battery Life | 210 Hours with Rechargeable Battery |
| Connection Type | Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately) |
| Construction | Rugged Cast Aluminum Housing, Stainless Steel Platform |
| Capacity | 3.00 kg |
| Display Type | 3 Window Backlit LCD, 3 Color Checkweighing LEDs |
| Readability | 1.000000 g |
| Linearity | 0.200000 g |
| Model | RC41M3-M |
| Repeatability | 0.200000 g |
| Temperature (Metric) Operating | -10°C to +40°C |
| Show More |