Learn More
Brannan™ Range Finder Hydrometers
GREENER_CHOICE
Product Code. 12861428
Click to view available options
Baume Range:
1 to 1.50
1.50 to 2
Unit Size:
Each
Product Code. 12861428
Brand: Brannan™ 10/532/0
Only null left
Add to basket
Add to basket
Brannan Range Finder Hydrometers, Baume Range: 1.50 to 2, Length: 250mm, Certifications/Compliance: UKAS, Quantity: 1Each, Subdivisions: 0.005g/mL
Specifications
Quantity 1Each
Type Range Finder Hydrometer
Increments 0.005 g/mL
Certifications/Compliance UKAS
Length (Metric) 250 mm
Baume Range 1.50 to 2
For Use With (Application) Specific Gravity Measurement
Specific Gravity Range 1.5 to 2