Learn More
Thermo Scientific™ Specialty Racks
Product Code. 10022573 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Holds:
Boxes (2 in.)
Boxes (3 in.)
General Freezer Items
Microplates
Microplates or Deepwell Microplates
Positions:
12 Boxes/Rack
15 Boxes/Rack
20 Boxes/Rack
22 Boxes/Rack
36 Plates/Rack
56 Plates/Rack
77 Plates/Rack
84 Plates/Rack
9 Boxes/Rack
98 microplates with lids, 119 microplates without lids, and 35 deepwell
Product Type:
Box Racks - Sliding Drawer
Side Access Freezer Rack (Adjustable)
Side Access Freezer Rack (Microplate/Deepwell)
Side Access Freezer Rack (Microplate/Locking Rod)
Specialty Rack - 10-Compartment Container
Specialty Rack - 8-Compartment Container
For Use With (Equipment):
13 cu. ft. Upright Freezers
17.3, 23 cu. ft. Upright Freezers
32 cu.ft. Revco Upright ULT
Thermo Scientific TDE and TSX Series (5 shelves)
missing translation for 'unitSize'
Each
Product Code. 10022573
missing translation for 'mfr': Thermo Scientific™ 920022
Only null left
Add to basket
Add to basket
Maximize the storage space in your ultra-low temperature freezer with these racks.
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
TRUSTED_SUSTAINABILITY
Specifications
Product Type Side Access Freezer Rack (Adjustable)
Color Silver
Holds Boxes (2 in.)
Material Stainless Steel
Positions 22 Boxes/Rack
Shelves 5
For Use With (Equipment) 17.3, 23 cu. ft. Upright Freezers
Width (Metric) 13.5 cm
Width (English) 5.3 in.
Height (Metric) 29.5 cm
Height (English) 11.6 in.
Depth (English) 24.6 in.
Depth (Metric) 62.5 cm