Learn More
Epredia™ RA Lamb Waxes and Embedding Media
Product Code. 12624077 Shop All Epredia Products
Click to view available options
Product Type:
Wax and Embedding Media
Quantity:
10 kg
1 x 10 kg
Unit Size:
10kg
Each
Product Code. 12624077
Brand: Epredia™ W1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Quickly and efficiently mount slides with Epredia™ RA Lamb waxing and embedding media for slide mounting.
Waxing and embedding media include:
  • Immersion oil
  • Embedding medium
  • Mounting medium
  • Chemical stain
  • Formaspill granules
  • Paraffin wax
  • Solution buffer
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wax and Embedding Media
Quantity 10 kg