Learn More
R&D Systems™ Human KA1/GRIK4 (NP_055434) VersaClone cDNA
Product Code. 18772638
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18772638
Brand: R&D Systems™ RDC0907
Glutamate Receptor KA1
The Human KA1/GRIK4 (NP_055434) VersaClone cDNA clone contains the complete ORF for KA1/GRIK4/Glutamate Receptor KA1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The KA1/GRIK4/Glutamate Receptor KA1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Gene hGRIK4
Accession Number NP_055434
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 2882bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans