Learn More
R&D Systems™ Human GABA-A R alpha 6 (NP_000802) VersaClone cDNA
gamma-Aminobutyric Acid Type A Receptor, alpha-6 Polypeptide
Brand:  R&D Systems™ RDC0827
Product Code. 18452200
  • € 322.00 / 10µg
Explore more special offers
Description
Description
The Human GABA-A R alpha 6 (NP_000802) VersaClone cDNA clone contains the complete ORF for GABA-AR alpha 6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GABA-AR alpha 6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hGABRA6
10 μg at 0.2 μg/μL
1375bp
Human
NP_000802
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans