missing translation for 'onlineSavingsMsg'
Learn More

(R)-Baclofen, Tocris Bioscience™

Product Code. 30001131
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective GABAB agonist; active enantiomer of (RS)-Baclofen (Cat. No. 0417)

More active enantiomer of (RS)-Baclofen, a selective GABAB agonist.

Racemate also available.

Chemical Identifiers

CAS 69308-37-8
Molecular Formula C10H12ClNO2
Molecular Weight (g/mol) 213.66
MDL Number MFCD01321057
InChI Key KPYSYYIEGFHWSV-UHFFFAOYNA-N
Synonym r-baclofen, arbaclofen, r-4-amino-3-4-chlorophenyl butanoic acid, d-baclofen, --baclofen, r---baclofen, 3r-4-amino-3-4-chlorophenyl butanoic acid, unii-nyu6utw25b
PubChem CID 44602
IUPAC Name 4-amino-3-(4-chlorophenyl)butanoic acid
SMILES NCC(CC(O)=O)C1=CC=C(Cl)C=C1

Specifications

Formula Weight 213.66
Percent Purity >99%
Quantity 10 mg
Chemical Name or Material (R)-Baclofen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'