missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-Amino-3-(2-bromophenyl)propionic acid, 98%, Thermo Scientific™ Product Code.: 15409545

(R)-3-Amino-3-(2-bromophenyl)propionic acid, 98%, Thermo Scientific™

Product Code. 15409545
250 mg
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15409545

Brand: Thermo Scientific Alfa Aesar H52044.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 737751-95-0
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD04113641
InChI Key OETRFEPZCAGEMK-MRVPVSSYSA-N
Synonym r-3-amino-3-2-bromophenyl propanoic acid, r-3-amino-3-2-bromo-phenyl-propionic acid, r-3-amino-3-2-bromophenyl propionic acid, 3r-3-amino-3-2-bromophenyl propanoic acid, s-3-amino-3-2-bromophenyl-propionic acid, h-d-b-phe 2-br-oh, r-2-bromo-beta-phe-oh, h-phg 2-br-c*ch2 oh, alphachiron 746639a1038Show MoreShow Less
PubChem CID 7046951
IUPAC Name (3R)-3-amino-3-(2-bromophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)C(CC(=O)O)N)Br

Specifications

Melting Point 232°C
Quantity 250 mg
Formula Weight 244.09
Optical Rotation +29° (c=1 in 0.5N NaOH)
Percent Purity 98%
Chemical Name or Material (R)-3-Amino-3-(2-bromophenyl)propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-Amino-3-(2-bromophenyl)propionic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.