Learn More
Quickfit™ Unbadged Joints Cones
Click to view available options
Cone Size:
10/19
12/21
14/23
19/26
24/29
29/32
34/35
40/38
45/40
55/44
7/16
Thickness (Metric) Wall:
1mm
1.5mm
1.8mm
2.00mm
2mm
2.30mm
5mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Pack of 50
Description
Free from any ceramic marking
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Manufactured to accuracy which surpasses requirements of ISO 383
- Pyrex™ borosilicate glass for resistance to chemical corrosion and thermal shock
- Cones manufactured with shank length of 100mm
Specifications
Specifications
| Type | Joint Cone |
| Cone Size | 24/29 |
| Thickness (Metric) Wall | 1.8mm |