Learn More
Quickfit™ Sealing Ring for Reaction Vessels
Silicone rubber sheathed with PTFE to provide a chemically resistant and grease free seal
Brand: SVL 296-90
58.90 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
This item is not returnable.
View return policy
Description
- For fitting between the flanges of SVL™ reaction vessels and lids
- Silicone rubber sheathed with PTFE to provide a chemically resistant and grease free seal
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Reaction vessel | |
Sealing ring | |
1PK |