Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
€ 36.10 - € 261.00
Specifications
For Use With (Equipment) | Dissimilar sized ground Glass joint |
---|---|
Includes | Small socket, large cone |
Material | Borosilicate Glass |
Product Type | Reduction adapter |
Product Code | Brand | Cone Size | Standard Taper Joint No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No. | Product Type | Price | Quantity & Availability | |||||
10203720
|
Quickfit
DA12 |
Cone Size: 19/26 | 14/23 | Reduction adapter |
€ 60.60
Pack of 2 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
10520681
|
Quickfit
DA13 |
Cone Size: 24/29 | 14/23 | Reduction adapter |
€ 55.60
Pack of 2 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
12634891
|
Quickfit
DA23 |
Cone Size: 24/29 | 19/26 | Reduction adapter |
€ 59.70
Pack of 2 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
12418876
|
Quickfit
DA14 |
Cone Size: 29/32 | 14/23 | Reduction adapter |
€ 68.20
Pack of 2 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
12520337
|
Quickfit
DA01 |
Cone Size: 14/23 | 10/19 | Reduction adapter |
€ 45.25
Pack of 2 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
10765861
|
Quickfit
DA24 |
Cone Size: 29/32 | 19/26 | Reduction adapter |
€ 75.70
Pack of 2 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
|||||
12448876
|
Quickfit
DA25 |
Cone Size: 34/35 | 19/26 | Reduction adapter |
€ 56.75
Pack of 2 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
12989253
|
Quickfit
DA35 |
Cone Size: 34/35 | 24/29 | Reduction adapter |
€ 60.25
Pack of 2 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
12654891
|
Quickfit
DA34 |
Cone Size: 29/32 | 24/29 | Reduction adapter |
€ 70.35
Pack of 2 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
12478876
|
Quickfit
DA45 |
Cone Size: 34/35 | 29/32 | Reduction adapter |
€ 64.55
Pack of 2 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
|||||
12050103
|
Quickfit
DA58 |
Cone Size: 50/42 | 34/35 | Reduction adapter |
€ 36.10
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
15212806
|
Quickfit
DA57 |
Cone Size: 45/40 | 34/35 | Reduction adapter |
€ 203.00
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
15597790
|
Quickfit
DA36 |
Cone Size: 40/38 | 24/29 | Reduction adapter |
€ 154.00
Pack of 5 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
15507800
|
Quickfit
DA37 |
Cone Size: 45/40 | 24/29 | Reduction adapter |
€ 201.00
Pack of 5 |
Estimated Shipment: 07-08-2025
Log in to see stock. |
|||||
15517800
|
Quickfit
DA38 |
Cone Size: 50/42 | 24/29 | Reduction adapter |
€ 196.00
Pack of 5 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
15527800
|
Quickfit
DA39 |
Cone Size: 55/44 | 24/29 | Reduction adapter | N/A | ||||||
15537800
|
Quickfit
DA47 |
Cone Size: 45/40 | 29/32 | Reduction adapter |
€ 261.00
Pack of 5 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
15547800
|
Quickfit
DA56 |
Cone Size: 40/38 | 34/35 | Reduction adapter |
€ 203.00
Pack of 5 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
- Useful for the connection of dissimilar sized ground glass joints
- Small sized socket at the top, large sized cone at the bottom
Specifications
Dissimilar sized ground Glass joint | |
Small socket, large cone | |
Borosilicate Glass | |
Reduction adapter |
Product Title
Quickfit™ Reduction Socket Adapter