Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
Brand:  Quickfit GB1/29
168.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15778017
  • € 182.00 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Description
  • Side arm to facilitate filling with mineral oil
  • Bubblers have capacity of approx 5ml
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reaction Vessel Bubbler
29/32