Learn More
Quickfit™ Multiple Glass Joint Adapter with Two Necks
Click to view available options
Cone Size:
19/26
Unit Size:
Each
Description
- With two necks :
- One vertical
- One angled at 45°
Specifications
Specifications
| Cone Size | 19/26 |
| Material | Borosilicate Glass |
| Product Type | Multiple neck adapter |