Learn More
Quickfit™ Borosilicate Glass Light Entrainer Receiver
Click to view available options
Type:
Light Entrainer Receiver
Light Entrainer Receiver with Stopcock
Capacity (Metric):
2 mL
7.5 mL
10 mL
25 mL
Unit Size:
Each
Beschreibung
- Complies with BS 756
- Dean and Stark pattern receivers for the determination of the water content in petroleum products
- For use with solvent entrainers which are lighter than water
- Borosilicate glass with graduated tube
Spezifikation
Spezifikation
| For Use With (Application) | For use with solvent entrainers which are lighter than water |
| Type | Light Entrainer Receiver |
| Material | Borosilicate Glass |
| Capacity | 2 mL |
| Certifications/Compliance | BS 756 |
| Capacity (Metric) | 2 mL |
| For Use With (Equipment) | Light entrainer |
| Size | Cone: 24/29, Socket: 19/26 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur