Learn More
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly
Complete kit including all glass components necessary to perform a solid-liquid extraction. Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly includes an extractor, condenser, and an evaporating flask.
Brand: Quickfit™ 2LRCSX
This item is not returnable.
View return policy
Description
- Each assembly comprises the basic components for solid-liquid extraction : Flask/Soxhlet extractor/Condenser/Associated lids and clips
- Choice of assemblies with different extractor capacities
Specifications
Soxhlet Extraction Assembly | |
Pyrex Borosilicate Glass | |
Ideal for solid-liquid extraction |
2000 mL | |
FR5L/5US Flask, EX5/105, DA5/100, JC100F Soxhlet extractor, C13/55 condenser, and associated lids and clip |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Complete Soxhlet Extractor Assembly > Capacity: 2000mL