Learn More
Hellma™ Quartz Suprasil Fluorescence Cells
Brand: Hellma™ 221-10-40
This item is not returnable.
View return policy
Specifications
Fluorescence Cell | |
Quartz Suprasil | |
1 Each | |
40 mm |
3500 μL | |
10mm | |
10 x 10 mm | |
12.5 mm |
Product Content Correction
Product Title
Hellma™ Quartz Suprasil Fluorescence Cells > Capacity: 3500μL; Height: 40mm