Learn More
Hellma™ Quartz Suprasil Fluorescence Cells
Brand:  Hellma™ 119004F-10-40
 View more versions of this product
Product Code. 10761493
Specifications
Fluorescence Cell
Quartz Suprasil
2/Pack
49.5 mm
Magnetic Stirrers
1500 μL
4mm
10 x 4 mm
12.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hellma™ Quartz Suprasil Fluorescence Cells > Capacity: 1500μL; Height: 49.5mm