missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Quantofix™ Chromate Semi-quantitative Test Strips
Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Violet
Unit Size:
50g
Description
Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
Specifications
| Color Indicator | White to Violet |
| Format | Strip |
| For Use With (Application) | Determination of Chromate |
| Graduations | 0, 3, 10, 30, 100mg/L CrO42- |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 30 sec. |
| Type | Semi-qualitative Test Strip |
Safety and Handling
Product Identifier
- QUANTOFIX Chromate
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction