Learn More
ValidR BV Qualification IQ-OQ-PQ
IQ-OQ-PQ ValidR a guide to process validation.
Brand:  ValidR BV BSC_IQ_OQ_PQ
Product Code. 30041483
  • € 140.00 / Each
Explore more special offers
Description
Description
  • IQ/OQ/PQ refers to the 3 activities that must be performed on equipment and machines as part of the validation of manufacturing processes: Installation Qualification (IQ), Operational Qualification (OQ), and Performance Qualification (PQ).
  • Evereything is installed correctly - IQ.
  • Everything is operating correctly, and that operating limits are understood - OQ.
  • It operates the way in which it is intended under load - PQ
Specifications
Specifications
Labconco PSM and BSC Cabinets
IQ/OQ Documentation