Learn More
BINDER™ Qualification Documents IQ/OQ
Product Code. p-7192910 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V
BINDER Chambers Series KBF P, KBF LQC, KBW, KBWF
BINDER Chambers Series KBF, KBF-S, KMF, MKF, MKFT
Product Type:
IQ/OQ Documentation (Hardcopy)
IQ/OQ Documentation (PDF)
Unit Size:
Each
Product Code. 16404019
Brand: BINDER™ 70570003
Qualification documents - IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
For Use With (Equipment) BINDER Chambers Series KBF P, KBF LQC, KBW, KBWF
Description Qualification documents - Digital in PDF format, IQ/OQ documents - supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, humidity, and light values
Duration On request
Product Type IQ/OQ Documentation (PDF)
Service Offering Supporting documents for validation performed by customers
Includes IQ/OQ checklists include calibration guide and comprehensive unit documentation