Learn More
WLD Tec™ Quadrupod Vierfu and Ceramic Protective Plate
Brand:  WLD Tec™ 6.000.930
Product Code. 11754169
  • € 59.00 / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Specifications
Quadrupod
155 mm
210 mm
155 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WLD Tec™ Quadrupod Vierfu and Ceramic Protective Plate > Height: 210mm