Learn More
Pyrido[2,3-b]pyrazine, 98%
Product Code. 11479877
Click to view available options
Quantity:
0.5 g
2.5 g
Unit Size:
0.5g
2.5g
Product Code. 11479877
Brand: Thermo Scientific Alfa Aesar B22843.88
Request Bulk or Custom Format
CAS: 322-46-3 | C7H5N3 | 131.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 322-46-3
Molecular Formula C7H5N3
Molecular Weight (g/mol) 131.14
MDL Number MFCD00006717
InChI Key YEYHFKBVNARCNE-UHFFFAOYSA-N
Synonym pyrido 2,3-b pyrazine, 1,4,5-triazanaphthalene, 5-azaquinoxaline, pyrido 2,3 pyrazine, pyrido 3,2-b pyrazine, acmc-20ahgw, 1,5-triazanaphthalene, pyridino 2,3-b pyrazine
PubChem CID 67580
IUPAC Name pyrido[2,3-b]pyrazine
SMILES C1=CN=C2N=CC=NC2=C1
Specifications
Melting Point 142°C to 144°C
Quantity 2.5 g
Formula Weight 131.14
Percent Purity 98%
Chemical Name or Material Pyrido[2,3-b]pyrazine