missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
2.5 L
Specifications
Specifications
Melting Point | -44°C |
CAS | 110-86-1 |
Assay Percent Range | min. 99.0% |
Molecular Formula | C5H5N |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
SMILES | C1=CC=NC=C1 |
IUPAC Name | pyridine |
Molecular Weight (g/mol) | 79.10 |
Density | 0.983 g/cm³ (20°C) |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction