Learn More
Pyrex™ Cylindrical Funnel with Semi-Micro Sintered Glass Disc
Ideal for filtering semi-micro quantities of material
Brand:  Quickfit SF4T31
111.15 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Product Code. 10737465
  • € 120.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Description
Ideal for filtering semi-micro quantities of material
  • Fitted with borosilicate glass sintered disc
  • Suitable for use with Quickfit™ MD range of receivers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
Quickfit™ MD: range of receivers
Sintered glass disc
Semi-micro Filter Funnel
14/23
10mm
3