Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Product Code. 15747957
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 15747957
Brand: Pyrex 3212/16WC
Only null left
Add to basket
Add to basket
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
  • Complies with ISO 4788
  • Calibrated to Class A tolerances
  • Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
  • Conformity H certified
  • Printed with individual serial number for batch certification
  • Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
  • Graduations in white enamel for improved visibility with dark colored solutions
  • Supplied with polyethylene stopper
TRUSTED_SUSTAINABILITY
Specifications
Base Type Hexagonal
Type Graduated Cylinder
Material Glass
Certifications/Compliance ISO 4788
Class A
Capacity (Metric) 1000 mL
Height (Metric) 505 mm
Includes Polyethylene Stopper
Stopper Number 45/40
Stopper Material Polyethylene