Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Product Code. p-8002541
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
29/32
Height (Metric):
107 mm
151 mm
160 mm
198 mm
Unit Size:
Each
Product Code. 12375263
Brand: Pyrex™ FD1L/4RE
Only null left
Add to basket
Add to basket
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 198 mm
Standard Taper Joint 29/32