Learn More
PYCR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55060-1-AP-150UL
 View more versions of this product
Product Code. 16826505
  • € 410.00 / 150µL
Description
The antibody is specific to PYCR2. It has no cross reaction to PYCR1.
PYCR2 belongs to the pyrroline-5-carboxylate reductase family. The function of the PYCR2 protein is not known.
Specifications
Specifications
PYCR2
Polyclonal
Unconjugated
PYCR2
P5C reductase 2, P5CR 2, P5CR2, PYCR2
Rabbit
Antigen Affinity Chromatography
RUO
29920
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.51 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96C36
PYCR2
Peptide
150 μL