Learn More
Lenz Laborglasinstrumente™ Pycnometers
Brand:  Lenz Laborglasinstrumente™ 05640037
 View more versions of this product
Product Code. 10080261
  • € 274.00 / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Specifications
1Each
Pycnometer
100 mL
Pycnometer
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Pycnometers > Capacity: 100mL; With Thermometer