Learn More
Kartell™ Plastilab™ PVC Non-toxic Isoflex Laboratory Hoses
Click to view available options
Diameter (Metric) Inner:
2 mm
4 mm
5 mm
6 mm
7 mm
8 mm
10 mm
Unit Size:
20m
Description
- Non-ageing and non-oxiding
- Resistant to both acids and alkalis
- Autoclavable
- Supplied in 20-meter rolls
- Packed in PE bags
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 8 mm |
| Material | PVC |
| Diameter (Metric) Outer | 12 mm |
| Overall Length (Metric) | 20 m |
| Thickness (Metric) | 2 mm |
| Packaging Quantity | 1 Pack |
| Color | Clear |