Learn More
OHAUS™ PVC Flask Clamp
Product Code. 15722519 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
1000 mL Flask
125 mL Flask
2000 mL Flask
250 mL Flask
500 mL Flask
Unit Size:
Each
Product Code. 15722519
Brand: OHAUS™ 30400100
Only null left
Add to basket
Add to basket
OHAUS™ PVC Flask Clamp is designed to securely grip and position various sizes of flasks.
  • Constructed of PVC
  • For use with flasks
  • Designed to securely hold flask in place
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1
Type Flask Clamp
Material PVC
For Use With (Equipment) 250 mL Flask
Description Flask Clamp PVC 250mL
Color Purple
Jaw Type Four Interlocking
Diameter (Metric) Inside 3.57 in