Learn More
Cole-Parmer™ PVC Check Valve, NPT(F)
Cole-Parmer™ PVC Check Valve, NPT(F) can be easily and quickly installed either vertically or horizontally.
Brand: Cole-Parmer™ 01341-00
Additional Details : Weight : 0.80000kg
Description
- Assembled with water soluble, silicone-free lubricant
- Constructed of a one-piece PVC body
Specifications
Check Valve | |
0.5 in. NPT Tubing | |
1 Pack |
PVC | |
Chemical processing or water/wastewater applications | |
60°C |