missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Purifier™ Class I Safety Enclosure

Provide your staff with personnel and environmental biohazard protection by outfitting your lab with the Labconco Purifier™ Class I Safety Enclosure.

Brand:  Labconco™ 3980321

7640.38 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17860052

  • € 7990.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Class I Safety Enclosure
37.08 in.
No Plug
94.1 cm
35.75 in.
50 Hz
72.74 cm
230 V
102 W
23.9 cm
Fluorescent
12 in.
90.8 cm
28.64 in.
230 V, 50 Hz
2 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Purifier™ Class I Safety Enclosure > 3 ft. width; 23.4 in. interior depth; 230V 50Hz, no plug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.