Learn More
Fisherbrand™ PTFE Wing Stir Bars
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
5.5 mm
8 mm
10 mm
13 mm
For Use With (Equipment):
11-12 mm diameter tubes
14-15 mm diameter tubes
6-7 mm diameter tubes
9-10 mm diameter tubes
Height (Metric):
9mm
10mm
11mm
16mm
20mm
Unit Size:
Pack of 10
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Alnico V magnet core |
| For Use With (Equipment) | 9-10 mm diameter tubes |
| Height (Metric) | 11mm |
| Diameter (Metric) | 8 mm |
| Shape | Wing |