Learn More
SI Analytics™ TZ 1798 PTFE Tube
Product Code. 10653982 Shop All SI Analytics Products
Click to view available options
For Use With (Equipment):
Titration Stand TM KF Connection, Moisture Bottle Reagent Bottle, Titration Stand, Titration Stand Waste Bottle
Unit Size:
Each
Product Code. 10653982
Brand: SI Analytics™ 285216882
Only null left
Add to basket
Add to basket
SI Analytics™ TZ 198 PTFE Tube is a meter long tube that is best suited for titration stand TM KF.
  • Suitable for titration stand TM KF and TM 235 KF
  • 1m long
  • For connection to titration vessel, reagent bottle, and waste bottle
TRUSTED_SUSTAINABILITY
Specifications
Type PTFE Tube
Material PTFE
For Use With (Equipment) Titration Stand TM KF Connection, Moisture Bottle Reagent Bottle, Titration Stand, Titration Stand Waste Bottle