Learn More
Kartell™ PTFE Tapered Stoppers
Quality PTFE constructed stopper to work in a variety of application. Kartell™ PTFE Tapered Stoppers can withstand high temperatures as well as a wide range of chemicals.
Brand: Kartell™ 0145500
This item is not returnable.
View return policy
Description
- Excellent chemical resistance
- Can withstand high temperatures
- Self-lubricating
- Suitable for foodstuff
Specifications
DIN Standards 12242 | |
Suitable for foodstuff | |
NS 14.5/23 Cone | |
12.2 mm | |
Tapered Stopper |
12.2mm | |
1/Pk. | |
Autoclavable | |
PTFE |