Learn More
Cowie Technology™ PTFE Straight Through Stopcocks
Shop All Fisherbrand Products
Click to view available options
Plug Bore:
2 mm
3 mm
4 mm
Unit Size:
Each
Description
- Medium vacuum pressure performance
- Unbreakable
- Chemically inert
- Grease free
- Supplied with borosilicate glass sidearms
Specifications
Specifications
| Type | Straight Through |
| Plug Bore | 4 mm |
| Description | Straight Through Stopcock |
| Material | PTFE |
| Autoclavable | Autoclavable |
| Fits Tubing ID | 10 mm |
| No. per Pack | 1 Each |
| Includes | Borosilicate glass sidearms |
| Plug Material | PTFE |