Learn More
IKA™ PTFE Slotted Bearing
For use with the IKA™ S 18 N - 19 G Dispersing Tool.
Brand:  IKA™ L004787
Product Code. 10760224
  • € 8.80 / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
  • Manufactured from PTFE
  • Aids in the homogenization process
  • Accessory item
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Slotted Bearing
PTFE
S 18 N - 19 G Dispersing Element
1 Ea.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ PTFE Slotted Bearing > For Use With: S 18N-19G Dispersing Element