Learn More
Glindemann™ PTFE Sealing Rings for taper joints (laboratory glassware)
For grease-free sealing of ground joints and plugs, extra tight and extra clean. The alternative to ground collar, grip collar sleeve and stopper made of PTFE.
POA - € 55.49
Product Code | Brand | dimProductType | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimProductType | Price | Quantity & Availability | |||||
10125930
|
Glindemann™
K7 |
€ 45.05
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||||
10268410
|
Glindemann™
K10 |
POA
Pack of 50 |
N/A | ||||||
10228460
|
Glindemann™
K12 |
€ 40.13
Pack of 50 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||||
10636521
|
Glindemann™
K14 |
€ 25.31
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10782131
|
Glindemann™
K19 |
POA
Pack of 50 |
N/A | ||||||
10646521
|
Glindemann™
K24 |
POA
Pack of 50 |
N/A | ||||||
10390001
|
Glindemann™
K29 |
€ 55.49
Pack of 50 |
In Stock
Log in to see stock. |
||||||
10257790
|
Glindemann™
K34 |
€ 55.23
Pack of 50 |
Estimated Shipment: 12-12-2024
Log in to see stock. |
||||||
10686901
|
Glindemann™
K45 |
POA
Pack of 50 |
N/A | ||||||
Description
- Air leak rate <10-8 to 10-6 mbar l/s (high vacuum, overpressure).
- Solvent leak rate in containers <0.1 to 0.03mg/d.
- Temperature resistant from -200 to +300°C.
- The elastic ring (0.07mm thick) adheres to the ground joint without a groove, is reusable and fits to ground joint clamps.
- Other sizes on request.
Specifications
PTFE |