Learn More
Aalborg™ PTFE N O-ring PTFE Frame
Product Code. 11728378
Click to view available options
For Use With (Equipment):
150 mm Gas/Liquid Flowmeter
Unit Size:
Each
Product Code. 11728378
Brand: Aalborg™ T11T2TA0
Only null left
Add to basket
Add to basket
Aalborg™ PTFE N O-ring PTFE Frame for 150mm T-style meter is for use with single tube flow meters, which are available with built in valves.
  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of Flow Tubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
  • Frame for T-meter
Specifications
For Use With (Equipment) 150 mm Gas/Liquid Flowmeter
Type Frame
Width (Metric) 150 mm