Learn More
DWK Life Sciences PTFE Hose Connection, with silicone seal (VMQ), complete with screw cap PBT, bent
For safe connection of hoses.
Brand:  DWK Life Sciences 286230802
113.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10030704
  • € 126.00 / Pack of 10
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Description
Hoses can be securely connected to laboratory apparatus using plastic nozzles screwed onto a glass thread using a screw cap with bore. Plastic nozzles made of PTFE for use with higher thermal and chemical resistance requirements.
  • Hose olives enable the connection of hoses to laboratory apparatuses
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Bent
10.5 mm
GL 18