Learn More
Horst™ PTFE HBSI Model Hard-Wearing Heating Tape
Brand:  Horst™ 20602
 View more versions of this product
Product Code. 10749812
  • € 194.00 / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Specifications
1 Lm
1 m
HBSI
1 Lm
∼230V Voltage
Hard-wearing Heating Tape
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Horst™ PTFE HBSI Model Hard-Wearing Heating Tape > Length: 1m