Learn More
Wheaton™ PTFE/Butadiene Septa
Brand:  Wheaton™ W240593
Product Code. 10378953
  • € 76.10 / Pack of 100
Estimated Shipment: 12-09-2025
to see stock.
Explore more special offers
Specifications
13 mm
Autoclavable
Septa
PTFE, Butadiene
100 Pk.
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Wheaton™ PTFE/Butadiene Septa > Outer Diameter: 13mm