Learn More
Fisherbrand™ PTFE Bottle Pourer
Product Code. 16236031 Shop All Fisherbrand Products
Click to view available options
Thread Size (Metric):
GL 30
GL 32
GL 38
GL 45
Unit Size:
Each
Product Code. 16236031
Only null left
Add to basket
Add to basket
Fisherbrand™ PTFE Bottle Pourer permits direct pouring of all liquids from reagent bottles in a safe and consistent manner.
  • Totally inert with PTFE encapsulated viton neck seal
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Description Fisherbrand™ PTFE Bottle Pourer
Material PTFE
Quantity 1/Ea.
Thread Size (Metric) GL 38
For Use With (Equipment) Reagent Bottles