Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Brand:  Fisherbrand™ 005.916150
 View more versions of this product
Product Code. 16255931
  • € 163.00 / Each
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
16 mm
-200°C to +280°C
150 mm
1