Learn More
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrers are for use with plain glass, metal or PTFE Shafts.
Brand: Fisherbrand™ 005.91075
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Adjustable Turbine Shaft Stirrer | |
10 mm | |
-200°C to +280°C |
75 mm | |
1 |