Learn More
PSI-6206, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
Specifications
Specifications
| Chemical Name or Material | PSI-6206 |
| CAS | 863329-66-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H13FN2O5 |
| Quantity | 100 mg |
| Synonym | RO 2433 GS-331007 |
| Solubility Information | DMSO : 100 mg/mL (384.29 mM; Need ultrasonic) |
| SMILES | O=C(NC1=O)C=CN1[C@H]2[C@](F)([C@@H]([C@H](O2)CO)O)C |
| Molecular Weight (g/mol) | 260.22 |
| Show More |
Research purposes only