Learn More
PSI-6206, MedChemExpress
Product Code. 16426388
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16426388
Brand: Medchem Express HY15236
MedChemExpress PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
PSI-6206 (RO 2433) is the deaminated derivative of PSI-6130, which is a potent and selective inhibitor of HCV NS5B polymerase. PSI-6206 low potently inhibits HCV replicon with EC90 of >100 μM.
Specifications
Chemical Name or Material PSI-6206
CAS 863329-66-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H13FN2O5
Quantity 10 mg
Synonym RO 2433 GS-331007
Solubility Information DMSO : 100 mg/mL (384.29 mM; Need ultrasonic)
SMILES O=C(NC1=O)C=CN1[C@H]2[C@](F)([C@@H]([C@H](O2)CO)O)C
Molecular Weight (g/mol) 260.22
Formula Weight 260.22
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only