Learn More
PSI-6130, MedChemExpress
MedChemExpress PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.
Brand:  Medchem Express HY-10165
Encompass
 View more versions of this product
Product Code. 16460056
  • € 352.50 / 2mg
Explore more special offers
Description
Description
PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.
Specifications
Specifications
PSI-6130
Off-White
C10H14FN3O4
R 1656
O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2O)F)O[C@@H]2CO)N
259.23
Research
COVID-19-anti-virus
817204-33-4
Research
2 mg
DMSO : 25 mg/mL (96.44 mM; Need ultrasonic)
259.23
95.0%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research purposes only