Learn More
PSI-6130, MedChemExpress
MedChemExpress PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.
Brand: Medchem Express HY-10165
This item is not returnable.
View return policy
Description
PSI-6130 is a potent and selective inhibitor of HCV NS5B polymerase, and inhibits HCV replication with a mean IC50 of 0.6 μM.Specifications
PSI-6130 | |
Off-White | |
C10H14FN3O4 | |
R 1656 | |
O=C1N=C(C=CN1[C@@H]([C@@](C)([C@@H]2O)F)O[C@@H]2CO)N | |
259.23 | |
Research | |
COVID-19-anti-virus |
817204-33-4 | |
Research | |
2 mg | |
DMSO : 25 mg/mL (96.44 mM; Need ultrasonic) | |
259.23 | |
95.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only